Learn More
Hirschmann™ Class A Volumetric Flasks, Amber Glass
Description
- With NS ground and hollow amber glass stopper
- Acc. to DIN EN ISO 1042
- Corrosion-free and resistant to thermal shock
- Conformity certified, with dated batch identification
Specifications
Specifications
| Certifications/Compliance | DIN EN ISO 1042 |
| Closure Color | Brown |
| Closure Material | Glass |
| Color | Brown |
| Capacity (Metric) | 20 mL |
| Material | Borosilicate Glass, Duran Glass |
| Height (Metric) | 110 mm |
| Limit of Error | ±0.04 mL |
| Standard Taper Joint | 12/21 NS Standard Ground |
| Tolerance | ±0.04 mL |
| Show More |