Learn More
Hirschmann™ Class A Volumetric Flasks, Amber Glass
Product Code. 10116590
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
150 mL
200 mL
500 mL
1000 mL
2000 mL
Standard Taper Joint:
10/19 NS Standard Ground
12/21 NS Standard Ground
14/23 NS Standard Ground
19/26 NS Standard Ground
24/29 NS Standard Ground
29/32 NS Standard Ground
7/16 NS Standard Ground
Unit Size:
Each
Pack of 2
Product Code. Capacity (Metric) Standard Taper Joint unitSize
10116590 1000 mL 24/29 NS Standard Ground Each
11859050 100 mL 12/21 NS Standard Ground Pack of 2
11786722 100 mL 14/23 NS Standard Ground Pack of 2
11878891 10 mL 10/19 NS Standard Ground Pack of 2
10761041 2000 mL 29/32 NS Standard Ground Each
11756722 10 mL 7/16 NS Standard Ground Pack of 2
11766722 20 mL 10/19 NS Standard Ground Pack of 2
11776722 25 mL 10/19 NS Standard Ground Pack of 2
11716742 200 mL 14/23 NS Standard Ground Pack of 2
11682229 20 mL 12/21 NS Standard Ground Pack of 2
11736742 500 mL 19/26 NS Standard Ground Pack of 2
11706742 50 mL 12/21 NS Standard Ground Pack of 2
11839600 50 mL 14/23 NS Standard Ground Pack of 2
13187939 5 mL 10/19 NS Standard Ground Pack of 2
30090420 150 mL 14/23 NS Standard Ground Pack of 2
15 options
Product Code. 10116590
Brand: Hirschmann™ 2640293
Only null left
Add to basket
Add to basket
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks, Amber Glass are manufactured from durable DURAN™ glass tubing.
  • With NS ground and hollow amber glass stopper
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Certifications/Compliance DIN EN ISO 1042
Closure Color Brown
Closure Material Glass
Color Brown
Capacity (Metric) 1000 mL
Material Borosilicate Glass, Duran Glass
Height (Metric) 300 mm
Limit of Error ±0.4 mL
Standard Taper Joint 24/29 NS Standard Ground
Tolerance ±0.4 mL
Graduated Yes
Class A