missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol Blue, Supelco™

Product Code. 15884320
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15884320 25 g 25g
15874320 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15884320 Supplier Supelco Supplier No. 1030260025
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Indicator ACS,Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-59-5
Grade Research
Molecular Weight (g/mol) 624.38

Specifications

MDL Number MFCD00005872
Synonym Bromothymol blue, 3′,3′′-Dibromothymolsulfonphthalein, BTB, Bromothymol blue
Quantity 25 g
Chemical Name or Material Bromothymol Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.