missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromostyrene, 95%, stabilized

Product Code. 10095153 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10095153

Brand: Thermo Scientific Acros 365801000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-81-7 | C8H7Br | 183.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-81-7
Molecular Formula C8H7Br
Molecular Weight (g/mol) 183.05
MDL Number MFCD00012229
InChI Key SRXJYTZCORKVNA-UHFFFAOYSA-N
Synonym 1-bromovinyl benzene, alpha-bromostyrene, benzene, 1-bromoethenyl, 1-bromostyrene, styrene, alpha-bromo, 1-phenylvinyl bromide, alpha-bromostyrol russian, styrene, .alpha.-bromo, unii-lf0sj1821n, alpha-bromostyrol
PubChem CID 66828
IUPAC Name 1-bromoethenylbenzene
SMILES BrC(=C)C1=CC=CC=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point -44°C
Color Colorless to Yellow
Density 1.4100g/mL
Boiling Point 67°C to 70°C (4.0 mmHg)
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C(Br)=CH2
Refractive Index 1.5870 to 1.589
Quantity 100 g
Specific Gravity 1.41
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alkanes,methylene chloride,ethanol,,2-propanol,acetone
Stabilizer about 0.05% 3, 5-di-tert-butylcatechol
Formula Weight 183.05
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material α-Bromostyrene, Stabilized
Show More Show Less
compliance-icons
Product Identifier
  • a-Bromostyrene, stabilized
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.