Learn More
Bromophenol Blue Reagent, Reagecon™
BIOPHARMA
Product Code. 12613623 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
100mL
Product Code. Quantity unitSize
12613623 1 Ea. 100mL
1 options
Product Code. 12613623
Brand: Reagecon 1012801
Specifications
Color Blue
Quantity 1 Ea.
compliance-icons compliance-icons
Product Identifier
  • bromophenol blue solution
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol