Description
Bromophenol Blue, Pure, Indicator Grade, Solid
Specifications
Specifications
| Melting Point | 279°C |
| CAS | 115-39-9 |
| Molecular Formula | C19H10Br4O5S |
| MDL Number | 5875 |
| Formula Weight | 669.96g/mol |
| Packaging | Glass Jar |
| Color | Red to Violet |
| Quantity | 5 g |
| Physical Form | Solid |