Learn More
Bromocresol Purple, Pure, Indicator Grade, Solid, Fisher Chemical™
Product Code. p-7072871 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10468590 5 g 5g
10030070 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10468590 Supplier Fisher Chemical Supplier No. B/4330/44
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
C21H16Br2O5S, CAS Number-115-40-2, 25g, Red, 540.21g/mol, 245 deg.C, 11681, Glass Jar, Powder Solid
TRUSTED_SUSTAINABILITY
Specifications
Melting Point 245°C
CAS 115-40-2
Molecular Formula C21H16Br2O5S
MDL Number 11681
Formula Weight 540.21g/mol
Packaging Glass Jar
Color Red
Quantity 5 g
Physical Form Powder