Learn More
Bromoacetic-1,2-13C2 Acid, TRC
Product Code. 30012192
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012192 1 g 1g
30012603 250 mg 250mg
30006314 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30012192 Supplier Toronto Research Chemicals Supplier No. TRCB6790781G
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 52947-00-9
Molecular Formula 13C2 H3 Br O2
Molecular Weight (g/mol) 140.93
Synonym Acetic-13C2 acid, bromo- (9CI), [1,2-13C2]Bromoacetic acid, Bromoacetic-1,2-13C acid, 2-Bromoacetic-1,2-13C2 acid, Bromoacetic Acid-13C2
IUPAC Name 2-bromoacetic acid
SMILES O[13C](=O)[13CH2]Br
Specifications
Quantity 1 g
InChI Formula InChI=1S/C2H3BrO2/c3-1-2(4)5/h1H2,(H,4,5)/i1+1,2+1
Formula Weight 139.9384
Chemical Name or Material Bromoacetic acid-13C2