Learn More
Bromine, Supelco™
Description
99.9999 Suprapur™
Specifications
Specifications
| Boiling Point | 58.8°C/1013 hPa |
| Melting Point | -7.3°C |
| CAS | 7726-95-6 |
| Quantity | 250 mL |
| MDL Number | MFCD00010896 |
| Synonym | Bromine, Bromine |
| Molecular Weight (g/mol) | 159.81 |
| Grade | Research |
| Density | 3.12 g/cm3 at 20°C (Liquid) |