Learn More
Bromindione, MedChemExpress
Product Code. 16498506
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498506 100 mg 100mg
16444507 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16498506 Supplier Medchem Express Supplier No. HYB0917
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
MedChemExpress Bromindione is a potent, long-acting, inandione-derived, oral anticoagulant compound.
Bromindione is a potent, long-acting, inandione-derived, oral anticoagulant compound.
Specifications
Chemical Name or Material Bromindione
CAS 1146-98-1
Color Brown
Purity Grade Notes Research
Molecular Formula C15H9BrO2
Quantity 100 mg
Synonym Fluidane Halinone
Solubility Information DMSO : 250 mg/mL (830.21 mM; Need ultrasonic)
SMILES O=C1C(C2=CC=C(Br)C=C2)C(C3=C1C=CC=C3)=O
Molecular Weight (g/mol) 301.13
Formula Weight 301.13
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation