Learn More
Bromide (from KBr) Standard: Br- @ 1000 μg/mL in H2O, VHG™
Product Code. 17987134
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17987134 100 mL 100mL
17989054 500 mL 500mL
17979054 250 mL 250mL
17979134 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17987134 Supplier VHG Labs Supplier No. VHGIBR100
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Bromide (from KBr) Standard: Br- @ 1000 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Bromide
CAS 24959-67-9
Methods Ion Chromatography
Analyte or Component Names Bromide
Shipping Condition Room Temperature
Molecular Formula Br-
Certifications/Compliance ISO 17034
Synonym Br−
SMILES [Br-]
IUPAC Name Bromide
Solvent or Matrix Water
Molecular Weight (g/mol) 79.9
Quantity 100 mL
Formula Weight 79.9 g/mol