Learn More
Usbeck™ Broad Base Stainless Steel Spatula Spoon
Product Code. 10483101
Change view
Click to view available options
Length (Metric):
120 mm
180 mm
210 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
10483101 180 mm Each
10620483 120 mm Each
10147391 210 mm Each
3 options
Product Code. 10483101
Brand: Usbeck™ 3333
Only null left
Add to basket
Add to basket
Scoop, mix and transfer substances with simplicity. Usbeck™ Broad Base Stainless Steel Spatula Spoon is ideal for scooping, mixing and transferring substances.
  • Stainless steel
  • Width of spoon 29mm
Specifications
Product Type Spoon Spatula
Width (Metric) 30 mm
Material Stainless Steel
Length (Metric) 180 mm