missing translation for 'onlineSavingsMsg'
Learn More

BRL-50481, MedChemExpress

Product Code. p-200098607
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484308 10 mg 10mg
16477027 5 mg 5mg
16475226 100 mg 100mg
16460637 50 mg 50mg
16433036 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16484308 Supplier Medchem Express Supplier No. HY109586

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BRL-50481 is a novel and selective inhibitor of PDE7 with IC50s of 0.15, 12.1, 62 and 490 μM for PDE7A, PDE7B, PDE4 and PDE3, respectively.

BRL-50481 is a novel and selective inhibitor of PDE7 with IC50s of 0.15, 12.1, 62 and 490 μM for PDE7A, PDE7B, PDE4 and PDE3, respectively.

Specifications

Chemical Name or Material BRL-50481
CAS 433695-36-4
Color White
Purity Grade Notes Research
Molecular Formula C9H12N2O4S
Quantity 10 mg
Solubility Information DMSO : 300 mg/mL (1228.15 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=S(C1=CC([N+]([O-])=O)=CC=C1C)(N(C)C)=O
Molecular Weight (g/mol) 244.27
Formula Weight 244.27
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.