Learn More
BRL-50481, MedChemExpress
Product Code. 16484308
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16484308
Brand: Medchem Express HY109586
MedChemExpress BRL-50481 is a novel and selective inhibitor of PDE7 with IC50s of 0.15, 12.1, 62 and 490 μM for PDE7A, PDE7B, PDE4 and PDE3, respectively.
BRL-50481 is a novel and selective inhibitor of PDE7 with IC50s of 0.15, 12.1, 62 and 490 μM for PDE7A, PDE7B, PDE4 and PDE3, respectively.
Specifications
Chemical Name or Material BRL-50481
CAS 433695-36-4
Color White
Purity Grade Notes Research
Molecular Formula C9H12N2O4S
Quantity 10 mg
Solubility Information DMSO : 300 mg/mL (1228.15 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=S(C1=CC([N+]([O-])=O)=CC=C1C)(N(C)C)=O
Molecular Weight (g/mol) 244.27
Formula Weight 244.27
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.