missing translation for 'onlineSavingsMsg'
Learn More

BRL-15572 dihydrochloride, MedChemExpress

Product Code. p-200101776
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16435548

Brand: Medchem Express HY13200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BRL-15572 dihydrochloride is a selective antagonist of h5-HT1D, displays high affinity for h5-HT1D receptors. BRL-15572 dihydrochloride could be useful pharmacological agents to characterise 5-HT1D receptor mediated responses.

BRL-15572 dihydrochloride is a selective antagonist of h5-HT1D, displays high affinity for h5-HT1D receptors. BRL-15572 dihydrochloride could be useful pharmacological agents to characterise 5-HT1D receptor mediated responses.

Specifications

Chemical Name or Material BRL-15572 dihydrochloride
Physical Form Solid
CAS 193611-72-2
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C25H29Cl3N2O
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 250 mg/mL (520.97 mM; Need ultrasonic) ∣H2O : 2 mg/mL (4.17 mM; ultrasonic and warming and heat to 60°C)
SMILES OC(CN1CCN(C2=CC(Cl)=CC=C2)CC1)C(C3=CC=CC=C3)C4=CC=CC=C4.[H]Cl.[H]Cl
Molecular Weight (g/mol) 479.87
Formula Weight 479.87
Percent Purity 99.77%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.