missing translation for 'onlineSavingsMsg'
Learn More

Brilliant Black BN, MP Biomedicals™

Product Code. p-4679911 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 11477972

Brand: MP Biomedicals 0215263305

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 2519-30-4
Molecular Formula C28H17N5Na4O14S4
Molecular Weight (g/mol) 867.664
InChI Key AGKKBBSOKGLVTM-YENKNCRGSA-J
Synonym CI 28440, Food Black 1
PubChem CID 6321379
IUPAC Name tetrasodium;(6E)-4-acetamido-5-oxo-6-[[7-sulfonato-4-[(4-sulfonatophenyl)diazenyl]naphthalen-1-yl]hydrazinylidene]naphthalene-1,7-disulfonate
SMILES CC(=O)NC1=C2C(=C(C=C1)S(=O)(=O)[O-])C=C(C(=NNC3=C4C=C(C=CC4=C(C=C3)N=NC5=CC=C(C=C5)S(=O)(=O)[O-])S(=O)(=O)[O-])C2=O)S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Formula Weight 867.7
Quantity 5 g
Physical Form Powder
Chemical Name or Material Brilliant Black BN
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.