Learn More
Invitrogen™ BrightComp eBeads™ Compensation Bead Kits
Product Code. p-7241185
Click to view available options
Description:
CFP BrightComp eBeads Compensation Bead Kit
GFP BrightComp eBeads Compensation Bead Kit
RFP BrightComp eBeads Compensation Bead Kit
YFP BrightComp eBeads Compensation Bead Kit
mCherry BrightComp eBeads Compensation Bead Kit
Unit Size:
Pack of 25
Product Code. 17897061
Brand: Invitrogen™ A54740
Edge
BrightComp eBeads compensation beads provide a consistent, accurate, and simple-to-use technique for setting flow cytometry compensation when using fluorescent protein-expressing samples.
• Three levels of intensity, perfect for any samples no matter the level of expression
• Near-identical, spectrally-matched dye to fluorescent protein
• Consistent, reliable performance
Accurate compensation is crucial for effective multicolor analysis in flow cytometry. Each kit provides polystyrene microspheres with near-identical, spectrally-matched fluorescent protein dye as well as negative control beads in a single dropper bottle.
TRUSTED_SUSTAINABILITY
Specifications
Description RFP BrightComp eBeads Compensation Bead Kit
Quantity 25 tests
Excitation Wavelength Range 561 nm
For Use With (Application) Flow Cytometry
Product Type Compensation Beads