Learn More
Invitrogen™ BrightComp eBeads™ Compensation Bead Kits
Product Code. 15980213
Change view
Click to view available options
Description:
CFP BrightComp eBeads Compensation Bead Kit
GFP BrightComp eBeads Compensation Bead Kit
RFP BrightComp eBeads Compensation Bead Kit
YFP BrightComp eBeads Compensation Bead Kit
mCherry BrightComp eBeads Compensation Bead Kit
Packungsgröße:
Pack of 25
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Description unitSize
15980213 GFP BrightComp eBeads Compensation Bead Kit Pack of 25
17897061 RFP BrightComp eBeads Compensation Bead Kit Pack of 25
17807071 YFP BrightComp eBeads Compensation Bead Kit Pack of 25
17817071 CFP BrightComp eBeads Compensation Bead Kit Pack of 25
17827071 mCherry BrightComp eBeads Compensation Bead Kit Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15980213 Lieferant Invitrogen™ Lieferanten-Nr. A10514
Edge
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
BrightComp eBeads compensation beads provide a consistent, accurate, and simple-to-use technique for setting flow cytometry compensation when using fluorescent protein-expressing samples.
  • Three levels of intensity, perfect for any samples no matter the level of expression
  • Near-identical, spectrally-matched dye to fluorescent protein
  • Consistent, reliable performance
Accurate compensation is crucial for effective multicolor analysis in flow cytometry. Each kit provides polystyrene microspheres with near-identical, spectrally-matched fluorescent protein dye as well as negative control beads in a single dropper bottle.
TRUSTED_SUSTAINABILITY
Spezifikation
Description GFP BrightComp eBeads Compensation Bead Kit
Content And Storage Store at 2°C to 8°C. DO NOT FREEZE.
For Use With (Equipment) Flow Cytometer
Quantity 25 tests
Color Green
Excitation Wavelength Range 488/525
Shipping Condition Room Temperature
For Use With (Application) Flow Cytometry
Product Line eBeads
Product Type Compensation Beads