missing translation for 'onlineSavingsMsg'
Learn More

Brequinar, MedChemExpress

Product Code. p-200096235
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16476897

Brand: Medchem Express HY108325

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Brequinar (DUP785) is a potent inhibitor of dihydroorotate dehydrogenase (DHODH) with an IC50 of 5.2 nM for human DHODH. Brequinar has potent activities against a broad spectrum of viruses. Brequinar also has an anti-SARS2 activity.

Brequinar (DUP785) is a potent inhibitor of dihydroorotate dehydrogenase (DHODH) with an IC50 of 5.2 nM for human DHODH. Brequinar has potent activities against a broad spectrum of viruses. Brequinar also has an anti-SARS2 activity.

Specifications

Chemical Name or Material Brequinar
Physical Form Solid
CAS 96187-53-0
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C23H15F2NO2
For Use With (Application) Cancer-Kinase/protease
Synonym DUP785 NSC 368390
Solubility Information DMSO : 25 mg/mL (66.60 mM; Need ultrasonic)
SMILES O=C(C1=C(C)C(C2=CC=C(C3=CC=CC=C3F)C=C2)=NC4=CC=C(F)C=C14)O
Molecular Weight (g/mol) 375.37
Formula Weight 375.37
Percent Purity 98.59%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.