Learn More
Brequinar, MedChemExpress
Product Code. 16476897
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16476897
Brand: Medchem Express HY108325
MedChemExpress Brequinar (DUP785) is a potent inhibitor of dihydroorotate dehydrogenase (DHODH) with an IC50 of 5.2 nM for human DHODH. Brequinar has potent activities against a broad spectrum of viruses. Brequinar also has an anti-SARS2 activity.
Brequinar (DUP785) is a potent inhibitor of dihydroorotate dehydrogenase (DHODH) with an IC50 of 5.2 nM for human DHODH. Brequinar has potent activities against a broad spectrum of viruses. Brequinar also has an anti-SARS2 activity.
Specifications
Chemical Name or Material Brequinar
Physical Form Solid
CAS 96187-53-0
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C23H15F2NO2
For Use With (Application) Cancer-Kinase/protease
Synonym DUP785 NSC 368390
Solubility Information DMSO : 25 mg/mL (66.60 mM; Need ultrasonic)
SMILES O=C(C1=C(C)C(C2=CC=C(C3=CC=CC=C3F)C=C2)=NC4=CC=C(F)C=C14)O
Molecular Weight (g/mol) 375.37
Formula Weight 375.37
Percent Purity 98.59%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.