missing translation for 'onlineSavingsMsg'
Learn More

BRD2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 22236-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16891775

  • 1281.92 NOK / 20µL
Estimated Shipment: 15-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a transcriptional regulator that belongs to the BET (bromodomains and extra terminal domain) family of proteins. This protein associates with transcription complexes and with acetylated chromatin during mitosis, and it selectively binds to the acetylated lysine-12 residue of histone H4 via its two bromodomains. The gene maps to the major histocompatability complex (MHC) class II region on chromosome 6p21.3, but sequence comparison suggests that the protein is not involved in the immune response. This gene has been implicated in juvenile myoclonic epilepsy, a common form of epilepsy that becomes apparent in adolescence. Multiple alternatively spliced variants have been described for this gene, but the full-length nature of some of these variants has not been determined.
Specifications

Specifications

BRD2
Polyclonal
Unconjugated
BRD2
BRD2, bromodomain containing 2, D6S113E, FSH, FSRG1, KIAA9001, NAT, O27.1.1, RING3, RNF3
Rabbit
Antigen Affinity Chromatography
RUO
6046
-20°C
Liquid
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence
0.81 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P25440
BRD2
BRD2 Fusion Protein Ag17589
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers