Learn More
Masterflex™ Brass Threaded Adapter
Description
- Available in a variety of configurations to meet a wide range of laboratory needs
- Adapts from a NPT male thread to a hose barb.
Specifications
Specifications
| Description | Barb x NPT(M) Connector |
| Material | Brass |
| Tubing Size | 1/2 in. |
| Hose Barb | 1/2 in. |
| Port Size | 1/2 in. NPT(M) |
| Thread Size | 1/2 in. NPT(M) |
| Product Type | Threaded Adapter |