Learn More
Cole-Parmer™ Brass Straight Compression Bulkhead Union
Product Code. 11825443
Click to view available options
Tubing Size:
1/4 in.
3/8 in.
Unit Size:
Each
Product Code. 11825443
Brand: Cole-Parmer™ 3141213
Only null left
Add to basket
Add to basket
Offers very good chemical resistance. Ideal when working with any rigid tubing, including tubing made from fluoropolymers, other rigid plastics and metals
  • Very good chemical resistance.
  • Compression fittings work well with rigid tubing such as PTFE or polypropylene.
  • A compression fitting has a nut that compresses rigid tubing against fitting body for high-pressure applications.
  • Compression connector family is ideal when working with rigid tubing.
  • When selecting your fittings, consider your tubing type, material compatibility and temperature and pressure conditions.
Specifications
Autoclavable Not Autoclavable
Material Brass
Min. Operating Temperature (English) -85°F
Min. Operating Temperature (Metric) -65°C
Tubing Size 1/4 in.
Maximum Pressure 400 psi
Max. Operating Temperature (English) 482°F
Max. Operating Temperature (Metric) 250°C
Product Type Compression Bulkhead Union