Learn More
Gas Arc™ Brass Gas Regulator
Product Code. 12910840
Encompass
Click to view available options
Diameter (Metric) Gauge:
3/8 mm
9.525 x 6.35 mm
9.525 x 9.525 mm
Connection Type:
Female L/H
Female R/H
G3/8 BSP L/H Brass Male
Unit Size:
Each
Product Code. 12910840
Brand: Gas Arc™ RSP00121
This item requires a price update prior to being ordered.
Request a Price for this item here.
Specifications
Connection Type G3/8 BSP L/H Brass Male
Type Gas Regulator
For Use With (Equipment) Cylinder
Material Brass
Pressure 300 bar (4350 PSI)
Thread Size 1/4 in. NPT Male
Diameter (Metric) Gauge 9.525 x 6.35 mm
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.