Learn More
Branson Ultrasonics™ SLP Cup Horn for Sonifiers SLPe, SLPt, 150E and 150T
Click to view available options
Length (English):
5 in.
Unit Size:
Each
Description
Enables sonification of very small volumes without horn coming into contact with sample
- Cup horn is attached to the inverted converter
- Inlet and outlet enables liquid to circulate within the chip horn, to reduce heat buildup and overflow
- Horn can also be used without liquid cooling circulation
- Process samples in complete isolation
- Ideal for infectious solutions
- Precludes possibility of any titanium residue
- Ideal for single or batch processing
- Ideal for very small volume processing
Cell Disruption, Liposome preparation, Protein shearing and extraction, Releasing cellular components, including DNA and RNA, Tissue culture Infectious solutions
Specifications
Specifications
| Diameter (English) | 3 in. |
| Length (English) | 5 in. |
| Material | Titanium |
| For Use With (Application) | Indirect method of disrupting |
| For Use With (Equipment) | Sonifier SLP, 150E, 150T |