Learn More
VACUUBRAND™ Rubber Tubing for Oil-Free Diaphragm Vacuum Pumps
Click to view available options
Overall Length (Metric):
3 m
Unit Size:
1m
Description
- Inner diameter 10 mm
- Three meter length
- Thick walls resist collapsing
- Made of high-quality caoutchouc rubber
Specifications
Specifications
| Autoclavable | Autoclavable |
| Diameter (English) Inner | 0.4 in. |
| Diameter (Metric) Inner | 11 mm |
| Material | Caoutchouc Rubber |
| Overall Length (English) | 9.8 ft. |
| Overall Length (Metric) | 3 m |
| For Use With | BrandTech pump models: ME1C, MEC, MZ1C, ME 2 and ME 4 NT Pumps |
| Packaging Quantity | 3-meter length |
| Color | Natural |
| Flexibility | Flexible |
| Show More |