Learn More
VACUUBRAND™ Quick Coupling for BVC Systems Made Prior to 2006
Product Code. 10088850
Click to view available options
For Use With:
BVC systems prior to 2006
Unit Size:
Each
Product Code. 10088850
Brand: VACUUBRAND™ 20688058
Only null left
Add to basket
Add to basket
Quick coupling for VACUUBRAND BVC21, BVC21B and BVC01 systems
  • For models made prior to 2006
  • Does not fit BVC basic, BVC control or BVC professional models
TRUSTED_SUSTAINABILITY
Specifications
For Use With BVC systems prior to 2006
Quantity 1 Ea.
Type Quick-coupling Aspirator