Learn More
BRAND™ Transferpettor™ Pipet Tips
Product Code. 10139900 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
1 to 5 mL
100 to 500 μL
2 to 10 mL
200 to 1000 μL
Length (English):
6.14 in.
6.2 in.
Length (Metric):
15.6 cm
16 cm
Color:
Green
Orange
Red
Yellow
Unit Size:
Pack of 10
Product Code. Volume (Metric) Length (English) Length (Metric) Color unitSize
10139900 200 to 1000 μL 6.14 in. 15.6 cm Yellow Pack of 10
10673191 1 to 5 mL 6.2 in. 16 cm Red Pack of 10
10393481 100 to 500 μL 6.2 in. 16 cm Green Pack of 10
10018790 2 to 10 mL 6.14 in. 15.6 cm Orange Pack of 10
4 options
Product Code. 10139900
Brand: BRAND™ 702854
Only null left
Add to basket
Add to basket
Reusable Positive Displacement Pipet Tips
  • Made of Polyethylene
  • Reusable Positive Displacement Pipet Tips
  • Compatible to use with BrandTech Transferpettor Positive Displacement Pipettes
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Color Yellow
For Use With Transferpette/Transferpette S
Material Polypropylene
Volume (Metric) 200 to 1000 μL
For Use With (Equipment) 200 to 1000μL Transferpettor
Length (English) 6.14 in.
Length (Metric) 15.6 cm