Learn More
BRAND™ Transferpette™ S Adjustable Single Channel Pipet Starter Sets
Shop All Brand Products
Click to view available options
Volume (Metric):
0.1 to 1, 0.5 to 10, 10 to 100 μL
0.5 to 10, 10 to 100, 100 to 1000 μL
100 to 1000, 500 to 5000, 1000 to 10,000 μL
2 to 20, 20 to 200, 100 to 1000 μL
Includes:
D-1, D-10, D-100 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes
D-10, D-100, D-1000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes
D-20, D-200, D-1000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes
D1000, D5000, D10000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes
Type:
Macro Set
Micro Set
Pipette Set, Standard
Starter Set, Adjustable
Unit Size:
Each
Beschreibung
Beinhaltet
Set contents:
- Three adjustable volume pipettes
- Three shelf/rack mounts
- Three filled polypropylene Tip Boxes
- Available as Standard Set, Micro Set, Midsize Set and Macro Set
Spezifikation
Spezifikation
| Autoclavable | Autoclavable |
| For Use With | Laboratory |
| Number of Channels | 1 |
| Product Line | Transferpette |
| Type | Starter Set, Adjustable |
| Volume (Metric) | 2 to 20, 20 to 200, 100 to 1000 μL |
| For Use With (Application) | Demanding laboratory applications |
| Includes | D-20, D-200, D-1000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur