Learn More
BRANDTECH™ VITLAB™ Square Narrow Mouth Bottles with Screw Caps
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
Closure Material:
LDPE
Material:
HDPE
Unit Size:
Pack of 12
Pack of 24
Description
- Bottles crafted from transparent high-density polyethylene
- Screw cap closures made of low-density polyethylene
- Space-saving design with a square cross-section and high shoulders
Specifications
Specifications
| Capacity | 3.38 oz., 100 mL |
| Capacity (Metric) | 100 mL |
| Capacity (English) | 3.38 oz. |
| Color | Transparent |
| Material | HDPE |
| Quantity | 24/Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Disposable | Single-use |
| Graduated | No |
| Show More |