Learn More
BrandTech™ Slide Staining Kit
Product Code. 10488036
Change view
Click to view available options
Dimensions (L x W x H):
100 x 87 x 51 mm
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
10488036 100 x 87 x 51 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10488036 Supplier BrandTech™ Supplier No. 99099
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
VITLAB POM Universal Slide Staining Kit.
  • Includes staining jack and staining rack
Specifications
For Use With (Equipment) Twenty-five 76 x 26 mm slides
Includes One staining jar and one staining rack
Dimensions (L x W x H) 100 x 87 x 51 mm