Learn More
BRAND™ Reagent Reservoirs
Product Code. 10347392 Shop All Brand Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Packaging:
40 Bags of 5
Individually Wrapped
Packs of 10
Unit Size:
Pack of 10
Pack of 100
Pack of 200
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Packaging unitSize
10347392 Sterile Individually Wrapped Pack of 100
10327202 Sterile 40 Bags of 5 Pack of 200
10063970 Non-sterile Packs of 10 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10347392 Supplier BRAND™ Supplier No. 703411
Only null left
Add to basket
Add to basket
Constructed of heavy-gauge polypropylene for long-lasting durability. Reagent Reservoirs are autoclavable and suitable for all 8- and 12-channel pipetters. Non-sterile version includes lid.
  • Heavy-gauge polypropylene
  • Suitable for all 8- and 12-channel pipetters
  • Autoclavable
  • Non-sterile version includes lid
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent
Capacity (Metric) 60 mL
Material Polypropylene
Sterility Sterile
Type Reagent Reservoir
For Use With (Application) For use with multichannel pipets. Without lid.
Packaging Individually Wrapped
Quantity 100/Pk.
Autoclavable Autoclavable
Channels 1
Disposable Limited-use
Bottom Shape V-bottom