Learn More
BRANDTECH™ VITLAB™ PFA Sample Containers
Product Code. 10060641
Change view
Click to view available options
Capacity (Metric):
60 mL
Capacity:
60 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Capacity Unit Size
10060641 60 mL 60 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10060641 Supplier BRANDTECH™ VITLAB™ Supplier No. 130397
  • ONLINE EXCLUSIVE PRICE
    730.00 NOK
    593.35 NOK / Pack of 1
    Save 136.65 NOK
    18% Off
 Restricted Item
Estimated Shipment: 15-12-2026
VITLAB PFA sample containers supplied with screw cap and are suitable for trace analysis work.
  • Suitable for sample collection, transport and storage.
  • Autoclavable at 121 C
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 60 mL
Product Type Sample Container
Capacity (English) 2.03 oz.
Material PFA
For Use With (Application) Sample Collection, Transport and Storage
Capacity 60 mL