Description
- For use with HandyStep™ S Repetitive Pipet
Specifications
Specifications
| Description | Shelf/rack mount |
| Autoclavable | Not Autoclavable |
| Color | Gray |
| Holds | Manual Pipettes |
| Material | Plastic |
| Positions | 1 |
| Shape | Circular |
| Stackable | No |
| Sterility | Non-sterile |
| For Use With (Equipment) | HandyStep S Electronic |
| Show More |