Learn More
BRAND™ Replacement Dispenser Cartridges
Replacement Cartridges for seripettor™ and seripettor™ pro, and QuickSip bottletop dispensers
Marque: BRAND™ 704502
354.69 NOK valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Informations supplémentaires : Weight : 0.02000kg
Description
- Sterile or non-sterile
- For use with seripettor™, seripettor™ pro and QuickSip bottletop dispensers
Spécification
Dispensing Cartridge | |
10 mL | |
Seripettor™ / Seripettor™ pro / QuikSip | |
Non-sterile |