Description
- Easily apply to plates and remove without the use of expensive equipment.
- Application with a hard rubber roller will ensure secure and uniform attachment of the film to the plate.
- Sealing films are available in different materials especially suited for storage, immunoassay, or cell and tissue culture:
- Polyethylene
- Polyester
- Aluminum
- Rayon
- Polypropylene
- Vinyl
Specifications
Specifications
| Color | Clear |
| Material | Vinyl |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| No. per Pack | 100 |
| Evaporation Barrier | No |
| For Use With (Application) | Automation |
| Pre-slit | Pre-punched |
| Product Type | Sealing Film |
| Adhesive | Pressure Sensitive |
| Show More |