Learn More
BRAND™ Self-adhesive Plate Sealing Film
Shop All Brand Products
Click to view available options
Material:
Aluminum
Polyester
Polyethylene, Polypropylene
Polypropylene
Rayon
Vinyl
For Use With (Application):
Automation
Cell and tissue culture
Cold storage
ELISA, PCR
ELISA, Real-time PCR
Luminescence measurements
Storage
Storage, fluorescence measurements
No. per Pack:
100
100 Sheets
50 Sheets
Unit Size:
Pack of 100
Pack of 50
Description
- Easily apply to plates and remove without the use of expensive equipment.
- Application with a hard rubber roller will ensure secure and uniform attachment of the film to the plate.
- Sealing films are available in different materials especially suited for storage, immunoassay, or cell and tissue culture:
- Polyethylene
- Polyester
- Aluminum
- Rayon
- Polypropylene
- Vinyl
Specifications
Specifications
| Color | White |
| Material | Polyethylene, Polypropylene |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| No. per Pack | 50 Sheets |
| Evaporation Barrier | Yes |
| For Use With (Application) | Automation |
| Pre-slit | No |
| Product Type | Sealing Film |
| Adhesive | Pressure Sensitive |
| Show More |