Learn More
BRAND™ BRANDplates immunoGrade™ 96-Well Microplates
Product Code. 10647143 Shop All Brand Products
Change view
Click to view available options
Color:
Black
Black, Clear Bottom
Clear
White
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round
Unit Size:
Pack of 100
Product Code. Color Well Shape unitSize
10647143 Clear Conical Bottom (V-well) Pack of 100
10193563 Clear Round Pack of 100
10032113 Clear Flat Bottom (F-well) Pack of 100
10477863 White Round Pack of 100
10438623 Black Round Pack of 100
10173853 Black, Clear Bottom Flat Bottom (F-well) Pack of 100
6 options
Product Code. 10647143
Brand: BRAND™ 781721
Only null left
Add to basket
Add to basket
An excellent choice for the majority of standard ELISAs as well as solid phase immunoassays
  • Optimized for the immobilization of IgG
  • High binding capacity for molecules with mixed hydrophobic and hydrophilic regions
  • Designed to minimize cross-contamination
  • Blue alphanumeric embossing for easy well identification on standard plates
  • Non-sterile
Order Info
Packaging is 20 stacks of 5 plates each (100 plates). Each stack comes with one lid.
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type High Binding
Color Clear
Product Type 96-well Microplate
Material Polystyrene
Volume (Metric) Well 330 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment immunoGrade
Total Volume (Metric) Well 360 μL
Well Shape Conical Bottom (V-well)
For Use With (Application) For the majority of standard ELISAs as well as solid phase immunoassays
Quantity 100/Cs.