Learn More
BRAND™ BLAUBRAND™ Glass Volumetric Flasks, Class A, USP, Certified
Product Code. 15282845 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
Closure Size:
NS 10/19
NS 12/21
NS 14/23
NS 19/26
NS 24/29
NS 29/32
Color:
Amber
Clear
Unit Size:
Each
Pack of 2
Product Code. Capacity (Metric) Closure Size Color unitSize
15282845 250 mL NS 14/23 Amber Pack of 2
15158434 5 mL NS 10/19 Clear Pack of 2
15192726 25 mL NS 10/19 Clear Pack of 2
15188264 50 mL NS 12/21 Clear Pack of 2
15178264 200 mL NS 14/23 Clear Pack of 2
15158264 250 mL NS 14/23 Clear Pack of 2
15891148 500 mL NS 19/26 Clear Pack of 2
15359332 1000 mL NS 24/29 Clear Each
15204069 2000 mL NS 29/32 Clear Each
15613967 5 mL NS 10/19 Amber Pack of 2
15623967 10 mL NS 10/19 Amber Pack of 2
15633967 20 mL NS 10/19 Amber Pack of 2
15262845 50 mL NS 12/21 Amber Pack of 2
15600327 200 mL NS 14/23 Amber Pack of 2
15643967 500 mL NS 19/26 Amber Pack of 2
15663967 1000 mL NS 24/29 Amber Each
15123129 20 mL NS 10/19 Clear Pack of 2
17 options
Product Code. 15282845
Brand: BRAND™ 37469
Indispensable for preparing dilutions and standard solutions
  • Manufactured from Borosilicate 3.3 glass, known for its low expansion coefficient and excellent chemical resistance
  • Available in transparent or amber glass
  • Glass stopper with drip-tip included
  • Featuring a full range of ISO size options, from 5 mL to 2000 mL
  • Calibrated 'To Contain'
  • Each unit is supplied with a batch number and a USP batch certificate in its original packaging for traceability
TRUSTED_SUSTAINABILITY
Specifications
Calibration To Contain
Certifications/Compliance DIN EN ISO 1042, USP batch certificate
Closure Material Glass
Color Amber
Capacity (Metric) 250 mL
Material Borosilicate Glass (3.3)
Limit of Error ±0.12 mL USP
No. per Case 2 Pieces
Product Type Volumetric Flask
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Mouth Style Narrow
Sterility Non-sterile
Class A
Bottom Shape Round
Closure Size NS 14/23