Learn More
BRAND™ BRANDplates™ Inlet Opening System PC Membrane Single Inserts
Shop All Brand Products
Click to view available options
Type:
PC Membrane 0.4 μm
PC Membrane 1.0 μm
PC Membrane 3.0 μm
PC Membrane 8.0 μm
Unit Size:
Pack of 48
Description
Specifications
Specifications
| Description | Inlet channels |
| Type | PC Membrane 3.0 μm |
| For Use With (Equipment) | 24-/6-well BRANDplates™ |
| Height (Metric) | 13 mm |
| Quantity | 48 |