Learn More
BRAND™ BRANDplates™ 384-Well Microplate
Shop All Brand Products
Click to view available options
Color:
White
Unit Size:
Pack of 5
Description
- Standard plate for the cultivation of adherent cell lines
- PS-surface with different chemical groups, such as carboxyl and hydroxyl groups, that are freely accessible
- Surface is hydrophilic compared with non-treated PS
- Serum components are easily bound onto the freely accessible chemical groups, allowing an indirect adhesion of cells
- Free from endotoxins, DNase, DNA, RNase, non-cytotoxic
- Sterile
- With lids
Specifications
Specifications
| Lid | With Lid |
| Color | White |
| Material | Polystyrene |
| No. of Wells | 384 |
| Sterility | Sterile |
| Surface Treatment | cellGrade |
| Total Volume (Metric) Well | 120 μL |
| Well Shape | Flat Bottom (F-well) |
| Dnase Free | Yes |
| RNase Free | Yes |
| Show More |