Learn More
Brady™ BradyPrinter i7100 Kit EU with Workstation PWID suite
Product Code. 16367595
Change view
Click to view available options
:
BradyPrinter i7100 EU; Color Capability: Monochrome; Print Speed: 150 mm/s; Print Technology: Direct Thermal, Thermal Transfer; Resolution: 600 dpi; Weight (Metric): 10 kg
Product Code.
16367595 BradyPrinter i7100 EU; Color Capability: Monochrome; Print Speed: 150 mm/s; Print Technology: Direct Thermal, Thermal Transfer; Resolution: 600 dpi; Weight (Metric): 10 kg
1 options
Product Code. 16367595
Brand: Brady™ 198613
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The BradyPrinter i7100 Industrial Label Printer is an accurate, heavy-duty printer that quickly prints on durable sleeves, printed circuit board labels and asset and component labels.
Produce a wide range of high quality identification materials with the BradyPrinter i7100 Kit for EU.
  • Powerful internal processor designed to handle up to 7000 labels per day
  • Intuitive color touch screen interface with distinctive icons to easily access printer menus
  • Optional I/O connection that allows printer to be triggered from programmable controller, foot pedal, sensor or other basic device
  • Center alignment of prints for high printing accuracy
  • Interchangeable platen rollers for optimized printhead lifetime
  • Includes Brady™ Workstation Product & Wire Identification Software Suite
Specifications
Display Type Color LCD
Print Technology Direct Thermal, Thermal Transfer
Width (English) 9.9 in.
Width (Metric) 252 mm
Height (English) 11.4 in.
Height (Metric) 290 mm
Depth (English) 18 in.
Depth (Metric) 460 mm
Autoclavable Not Autoclavable
Sterility Non-sterile
Color Capability Monochrome
Includes i7100 Printer, EU Power Cord, 2 Empty Ribbon Cores, USB Cable, Product CD with Driver, User Manual, Brady™ Workstation Product & Wire ID Suite
Languages Arabic, Bulgarian, Croatian, Czech, Danish, Dutch, English, Finnish, French, German, Greek, Hungarian, Italian, Lithuanian, Norwegian, Polish, Portuguese, Romanian, Russian, Serbian, Slovenian, Spanish, Swedish, Turkish
Memory 256 MB RAM, 50 MB IFFS Flash, SD Card, Slot Up To 512 GB
Model i7100
Print Speed 150 mm/s
Resolution 600 dpi
Connector Types Bluetooth™, Ethernet, I/O Connection, Serial, USB
Operating System Windows™ 10, Windows™ 7, Windows™ 8, Windows™ Servers 2012, 2012 R2, and 2016
Temperature Range (Metric) 0°C to 40°C
Weight (English) 22 lb.
Weight (Metric) 10 kg
For Use With (Application) Barcode and Inventory Labeling, Electrical Labeling, Laboratory Labeling
Warranty 2 Year
Voltage 110/240 V
Alternate Part Number i7100-600-EU-PWID