Learn More
Brady™ BradyPrinter i7100 - EU with Brady Workstation LAB Suite
Product Code. 17197011
Change view
Click to view available options
Resolution:
300 dpi
600 dpi
Description:
With Software, Peel Function
With Softwate
Unit Size:
1 set
Product Code. Resolution Description unitSize
17197011 300 dpi With Software, Peel Function 1 set
17177011 300 dpi With Softwate 1 set
17117021 600 dpi With Softwate 1 set
17137021 600 dpi With Software, Peel Function 1 set
4 options
Product Code. 17197011
Brand: Brady™ 306883
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Industrial thermal transfer benchtop printer is ideal for customers requiring reliable high volume printing on lots of different materials within the Aerospace, Defence and Mass Transit, Electronics, Automotive, Logistics, Electrical, Voice-Datacom and LAB markets.
  • Industrial thermal transfer benchtop printer
  • Ideal for customers requiring reliable high volume printing on lots of different materials within the following markets:
    • Aerospace
    • Defence and Mass Transit
    • Electronics
    • Automotive
    • Logistics
    • Electrical
    • Voice-Datacom
    • LAB
  • It guarantees accurate printing on sleeves, tags, labels, tapes, cable markers and much more
  • The printer comes with the Brady Workstation Laboratory Identification Software suite enabling you to immediately start designing professional labels
  • This suite combines frequently used software apps to create laboratory identification labels
Specifications
Product Type Industrial Printer
Display Type Color Touchscreen
Print Technology Thermal Transfer
Width (Metric) 252.00 mm
Height (Metric) 288.00 mm
Depth (Metric) 460.00 mm
Dimensions (L x W x H) 460.00 x 252.00 x 288.00 mm
Color Capability Monochrome
Description With Software, Peel Function
Includes Peel and Present module pre-installed. It ships with an EU power cord, 2 empty ribbon cores, a USB cable, a product CD holding the driver, the user manual in languages, the configuration/settings manual in English, a printed user manual in English and Brady Workstation Laboratory ID Suite
Print Speed 300.00 mm/s
Resolution 300 dpi
Software Type Brady Workstation LAB Suite
Power Supply Details AC Adaptor
Alternate Part Number i7100-300P-EU-LABS