Learn More
Brady™ BradyPrinter i7100 for EU
Product Code. 16379171
Click to view available options
:
BradyPrinter i7100 EU; Print Speed: 150 mm/s; Resolution: 300 dpi; Print Technology: Direct Thermal, Thermal Transfer; Weight (Metric): 10 kg; Color Capability: Monochrome
Product Code. 16379171
Brand: Brady™ 149046
The BradyPrinter i7100 Industrial Label Printer is an accurate, reliable, heavy-duty printer that quickly prints on durable sleeves, printed circuit board labels and asset and component labels.
The Brady™ i7100 Label Printer has a powerful internal processor designed to handle up to 7000 labels per day.
  • Intuitive color touch screen interface with distinctive icons to easily access the printer's menus
  • Optional I/O connection that allows your printer to be triggered from the programmable controller, foot pedal, sensor or other basic device
  • Center alignment of prints for high printing accuracy
  • Interchangeable platen rollers for optimized printhead lifetime
  • A wide range of high quality identification materials
  • Incudes QWERTY keyboard and free Brady™ Workstation Basic Software Suite
Specifications
Display Type Color LCD
Print Technology Direct Thermal, Thermal Transfer
Width (English) 9.9 in.
Width (Metric) 252 mm
Height (English) 11.4 in.
Height (Metric) 290 mm
Depth (English) 18 in.
Depth (Metric) 460 mm
Autoclavable Not Autoclavable
Sterility Non-sterile
Color Capability Monochrome
Includes i7100 Printer, EU Power Cord, 2 Empty Ribbon Cores, USB Cable, Product CD with Driver, User Manual, Brady™ Workstation Basic Suite
Languages Arabic, Bulgarian, Chinese (Tradditional), Chinese (Simplified), Croatian, Czech, Danish, Dutch, English, Finnish, French, German, Greek, Hungarian, Italian, Lithuanian, Norwegian, Polish, Portuguese, Romanian, Russian, Serbian, Slovenian, Spanish, Swedish, Turkish
Memory 256 MB RAM, 50 MB IFFS Flash, SD Card, Slot Up To 512 GB
Model i7100
Print Speed 150 mm/s
Resolution 300 dpi
Connector Types Bluetooth™, Ethernet, I/O Connection, Serial, USB
Operating System Windows™ 10, Windows™ 7, Windows™ 8, Windows™ Servers 2012, 2012 R2, and 2016
Temperature Range (Metric) 5°C to 40°C
Weight (English) 22 lb.
Weight (Metric) 10 kg
For Use With (Application) Circuit Board and Electrical Labeling, General Identification, Safety Identification
Warranty 2 Year
Voltage 110/240 V
Alternate Part Number i7100-300-EU