Learn More
Brady™ BradyJet™ Pre-Cut Vinyl Labels
Product Code. 16150491
Change view
Click to view available options
Dimensions (LxW):
3.35 x 2.17 in.
4.33 x 2.95 in.
6.10 x 4.13 in.
Unit Size:
Each
Product Code. Dimensions (LxW) unitSize
16150491 6.10 x 4.13 in. Each
16110491 3.35 x 2.17 in. Each
16130491 4.33 x 2.95 in. Each
3 options
Product Code. 16150491
Brand: Brady™ 150032
Only null left
Add to basket
Add to basket
Brady™ BradyJet™ Pre-Cut Vinyl Labels are ideal for a wide range of identification applications.
  • Constructed of indoor-use general purpose vinyl
  • Features a specially-formulated topcoat for improved durability and color
  • Specially-formulated topcoat enhances colors and label appearance
  • Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting
  • Topcoat protects print from smudging and harsh conditions
  • Inkjet-receptive labels for use with the BradyJet™ J2000 Color Label Printer
Specifications
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
Format Roll
Material Vinyl
Printable Sides Single-side Printable
Print Technology Ink Jet
Product Type Label
Sterility Non-sterile
For Use With (Application) Pipe and Valve Marking, Safety and Compliance ID, Visual Workplace Labeling, Warehouse Marking, Lockout Tagout Labeling, GHS and Chemical Labels, Arc Flash Labels
For Use With (Equipment) BradyJet™ J2000 Printer
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
No. per Pack 200 Labels/Bag
Quantity 200/Roll
Shape Rectangle
Temperature Range (English) -40°F to +140°F
Temperature Range (Metric) -40°C to +60°C
Thickness (English) 0.0053 in.
Thickness (Metric) 0.135 mm
Dimensions (LxW) 6.10 x 4.13 in.
Length (English) 6.10 in.
Width (English) 4.13 in.
Width (Metric) 104.90 mm
Height (English) 6.10 in.
Height (Metric) 154.94 mm
Diameter (English) 1.88 in. I.D.
Alternate Part Number J20-262-2595