Learn More
Brady™ BradyJet™ Polyester Tags
Brady™ BradyJet™ Polyester Tags are ideal for a wide range of identification applications and can be use on a variety of laboratory equipment.
Brand: Brady™ 150008
Additional Details : Weight : 0.45000kg
Description
- Ideal for use in general facility labeling, including accident prevention and confined space tags
- Compliant with OSHA standard 1910.147 for lockout tagout
- Compliant with OSHA standard 1910.145 for accident prevention tags
- Material is a thick polyester tagstock material, with an inkjet receptive vinyl adhered
Specifications
Non-adhesive | |
Matte | |
Pipe & Valve Marking; Safety & Compliance ID; Visual Workplace Labeling; Warehouse Marking; Lockout Tagout Labeling; GHS & Chemical Labels; Arc Flash Labels | |
−40 to 140°F | |
50 ft. x 3.25 in. |
White | |
Polyester | |
BradyJet™ J2000 Printer | |
-40°C to +60°C | |
1.88 in. I.D. |