Learn More
Brady™ BradyJet™ J2000 Repositionable Inkjet Vinyl Labels
The special aggressive adhesive on this Repositionable Inkjet Material stays in place when you need it to, but removes cleanly without leaving adhesive residue making it ideal for 5S and inventory applications.
Brand: Brady™ 151108
Additional Details : Weight : 0.55800kg
Description
- Durable and flexible material making it ideal for areas where identification is frequently changed
- Service temperature of -40 to 140°F (-40 to 60°C)
Specifications
Acrylic | |
Matte | |
Repositionable | |
BradyJet™ J2000 | |
-40°F to +140°F |
White | |
Vinyl | |
Temporary Labeling | |
1 | |
100 ft. x 2.25 in. |