Learn More
Brady™ Vinyl Tapes (B-595) for BMP71 Printer
Click to view available options
Color:
Black
Blue
Brown
Clear
Gold
Gray
Green
Light Blue
Ochre
Pink
Purple
Sky Blue
Tan
Unit Size:
Each
Description
- Vinyl film with permanent adhesive
- Adheres well to pipes, walls, equipment, windows - most clean, dry surfaces
- Resistant to grease, oil and most industrial chemicals
- Five year outdoor durability in temperatures from 82° to -40°C (180° to -40°F, can be applied as low as -18°C (0°F)
Specifications
Specifications
| Color | Ochre |
| Material | Vinyl (B-595) |
| Product Type | Tape |
| For Use With (Application) | Labeling pipes, walls, equipment, windows |
| For Use With (Equipment) | BMP™71 Printer |
| Quantity | 1 Ea. |
| Size (English) | 50 ft. x 1 in. |
| Dimensions (LxW) | 15.24 m x 2.54 cm (50 ft. x 1 in.) |
| Length (English) | 50 ft. |
| Length (Metric) | 15.24 m |
| Show More |