Learn More
Brady™ ToughStripe™ Labels with Overlaminate
Product Code. 11741016
Change view
Click to view available options
Color:
Black, Clear
Blue
Green, Clear
Orange, Clear
Red, Clear
White, Clear
Yellow, Clear
Length (English):
100 ft.
Width (English):
1.12 in.
2.25 in.
Width (Metric):
2.86 cm
5.71 cm
Unit Size:
1 roll
Product Code. Color Length (English) Width (English) Width (Metric) unitSize
11741016 Orange, Clear 100 ft. 1.12 in. 2.86 cm 1 roll
11721006 White, Clear 100 ft. 1.12 in. 2.86 cm 1 roll
11711006 White, Clear 100 ft. 2.25 in. 5.71 cm 1 roll
11731006 Black, Clear 100 ft. 2.25 in. 5.71 cm 1 roll
11741006 Black, Clear 100 ft. 1.12 in. 2.86 cm 1 roll
11751006 Yellow, Clear 100 ft. 2.25 in. 5.71 cm 1 roll
11761006 Yellow, Clear 100 ft. 1.12 in. 2.86 cm 1 roll
11781006 Red, Clear 100 ft. 1.12 in. 2.86 cm 1 roll
11771006 Red, Clear 100 ft. 2.25 in. 5.71 cm 1 roll
11791006 Blue 100 ft. 2.25 in. 5.71 cm 1 roll
11701016 Blue 100 ft. 1.12 in. 2.86 cm 1 roll
11721016 Green, Clear 100 ft. 1.12 in. 2.86 cm 1 roll
11731016 Orange, Clear 100 ft. 2.25 in. 5.71 cm 1 roll
11711016 Green, Clear 100 ft. 2.25 in. 5.71 cm 1 roll
14 options
Product Code. 11741016
Brand: Brady™ 142175
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
  • 1 roll of B-483 polyester
  • 1 roll of B-634 polyester overlaminate
Create custom-printed floor labels quickly and easily
Easy Three-Step Process
  • Print label
  • Position on floor
  • Apply overlaminate
Specifications
Certifications/Compliance RoHS 2002/95/EC
Description GlobalMark ToughStripe Floor Marking Tape with Overlaminate
Adhesive Type Rubber, Permanent
Coating None
Color Orange, Clear
Finish Gloss
Format Tape
Material Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) For high adhesion to textured metals and low surface energy Plastics
For Use With (Equipment) GlobalMark™ 2 and BMP™ 71 label printer
Includes 1 Roll of tape and 1 roll of over laminate
Labels per Row 1
Min. Operating Temperature (Metric) -18°C
No. per Pack 2 Rolls/Kit
Quantity 1 Kit
Temperature Range (English) 0°F to 130°F
Temperature Range (Metric) -18°C to +54°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.203 mm
Dimensions (LxW) 100 ft. x 1.12 in. (30.48 m x 2.86 cm)
Length (English) 100 ft.
Length (Metric) 30.48 m
Width (English) 1.12 in.
Width (Metric) 2.86 cm