Learn More
Brady™ ToughStripe™ Labels with Overlaminate
Product Code. 11711006
Change view
Click to view available options
Color:
Black, Clear
Blue
Green, Clear
Orange, Clear
Red, Clear
White, Clear
Yellow, Clear
Length (English):
100 ft.
Width (English):
1.12 in.
2.25 in.
Width (Metric):
2.86 cm
5.71 cm
Unit Size:
1 roll
Product Code. Color Length (English) Width (English) Width (Metric) unitSize
11711006 White, Clear 100 ft. 2.25 in. 5.71 cm 1 roll
11721006 White, Clear 100 ft. 1.12 in. 2.86 cm 1 roll
11731006 Black, Clear 100 ft. 2.25 in. 5.71 cm 1 roll
11741006 Black, Clear 100 ft. 1.12 in. 2.86 cm 1 roll
11751006 Yellow, Clear 100 ft. 2.25 in. 5.71 cm 1 roll
11761006 Yellow, Clear 100 ft. 1.12 in. 2.86 cm 1 roll
11781006 Red, Clear 100 ft. 1.12 in. 2.86 cm 1 roll
11771006 Red, Clear 100 ft. 2.25 in. 5.71 cm 1 roll
11791006 Blue 100 ft. 2.25 in. 5.71 cm 1 roll
11701016 Blue 100 ft. 1.12 in. 2.86 cm 1 roll
11721016 Green, Clear 100 ft. 1.12 in. 2.86 cm 1 roll
11731016 Orange, Clear 100 ft. 2.25 in. 5.71 cm 1 roll
11741016 Orange, Clear 100 ft. 1.12 in. 2.86 cm 1 roll
11711016 Green, Clear 100 ft. 2.25 in. 5.71 cm 1 roll
14 options
Product Code. 11711006
Brand: Brady™ 142154
Only null left
Add to basket
Add to basket
Includes
  • 1 roll of B-483 polyester
  • 1 roll of B-634 polyester overlaminate
Create custom-printed floor labels quickly and easily
Easy Three-Step Process
  • Print label
  • Position on floor
  • Apply overlaminate
Specifications
Certifications/Compliance RoHS 2002/95/EC
Description GlobalMark ToughStripe Floor Marking Tape with Overlaminate
Adhesive Type Rubber, Permanent
Coating None
Color White, Clear
Finish Gloss
Format Tape
Material Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) For high adhesion to textured metals and low surface energy Plastics
For Use With (Equipment) GlobalMark™ 2 and BMP™ 71 label printer
Includes 1 Roll of tape and 1 roll of over laminate
Labels per Row 1
Min. Operating Temperature (Metric) -18°C
No. per Pack 2 Rolls/Kit
Quantity 1 Kit
Temperature Range (English) 0°F to 130°F
Temperature Range (Metric) -18°C to +54°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.203 mm
Dimensions (LxW) 100 ft. x 2.25 in. (30.48 m x 5.71 cm)
Length (English) 100 ft.
Length (Metric) 30.48 m
Width (English) 2.25 in.
Width (Metric) 5.71 cm
Alternate Part Number GMK B-483 WT 57,2mm & B-634OLam
Show More Show Less
Product Title
Select an issue
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.